Assay Details
| Gene Symbol: SHISA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018067026.1 | XP_017922515.1 | 4 - 5 | 499 | 68 | |
| XM_018067027.1 | XP_017922516.1 | 3 - 4 | 449 | 68 | ||
| XM_018067028.1 | XP_017922517.1 | 2 - 3 | 249 | 68 | ||
| GenBank mRNA | JO542304.2 | - | 1 - 2 | 226 | 68 | |
More Information
Set Membership: |
|

SHISA5
Genomic Map