Assay Details
| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046416.1 | XP_017901905.1 | 3 - 4 | 324 | 61 | |
| XM_018046417.1 | XP_017901906.1 | 3 - 4 | 322 | 61 | ||
| XM_018046418.1 | XP_017901907.1 | 2 - 3 | 301 | 61 | ||
More Information
Set Membership: |
|

FCRLB
Genomic Map