Assay Details
| Gene Symbol: TMEM174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694561.3 | XP_005694618.3 | 1 - 2 | 710 | 86 | |
| GenBank mRNA | JO436781.2 | - | 1 - 2 | 752 | 86 | |
More Information
Set Membership: |
|

TMEM174
Genomic Map