Assay Details
| Gene Symbol: CALHM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684536.3 | XP_005684593.1 | 1 - 2 | 603 | 82 | |
| GenBank mRNA | KA319557.1 | - | 1 - 2 | 667 | 82 | |
More Information
Set Membership: |
|

CALHM4
Genomic Map