Assay Details
| Gene Symbol: KLK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692758.3 | XP_005692815.1 | 3 - 4 | 394 | 70 | |
| GenBank mRNA | KA352230.1 | - | 3 - 4 | 983 | 70 | |
More Information
Set Membership: |
|

KLK13
Genomic Map