Assay Details
| Gene Symbol: YIPF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700717.3 | XP_005700774.1 | 3 - 4 | 798 | 86 | |
| XM_013976422.2 | XP_013831876.1 | 3 - 4 | 802 | 86 | ||
More Information
Set Membership: |
|

YIPF6
Genomic Map