Assay Details
Gene Symbol: CD37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692708.3 | XP_005692765.2 | 6 - 7 | 821 | 64 | |
XM_018062768.1 | XP_017918257.1 | 6 - 7 | 821 | 64 | ||
XM_018062769.1 | XP_017918258.1 | 6 - 7 | 821 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
