Assay Details
| Gene Symbol: HTR2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700259.2 | XP_005700316.1 | 2 - 3 | 410 | 74 | |
| GenBank mRNA | JO619977.2 | - | 2 - 3 | 361 | 74 | |
More Information
Set Membership: |
|

HTR2C
Genomic Map