Assay Details
| Gene Symbol: LRRC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058765.1 | XP_017914254.1 | 2 - 3 | 1797 | 62 | |
| XM_018058766.1 | XP_017914255.1 | 2 - 3 | 1797 | 62 | ||
| XM_018058767.1 | XP_017914256.1 | 3 - 4 | 766 | 62 | ||
| XM_018058768.1 | XP_017914257.1 | 3 - 4 | 633 | 62 | ||
| XM_018058769.1 | XP_017914258.1 | 3 - 4 | 657 | 62 | ||
More Information
Set Membership: |
|

LRRC24
Genomic Map