Assay Details
| Gene Symbol: TMEM53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678555.3 | XP_005678612.2 | 2 - 3 | 338 | 91 | |
| GenBank mRNA | JO429936.2 | - | 2 - 3 | 389 | 91 | |
More Information
Set Membership: |
|

TMEM53
Genomic Map