Assay Details
| Gene Symbol: ETFBKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680696.3 | XP_005680753.1 | 3 - 4 | 831 | 90 | |
| GenBank mRNA | KA320740.1 | - | 1 - 2 | 138 | 90 | |
More Information
Set Membership: |
|

ETFBKMT
Genomic Map