Assay Details
| Gene Symbol: GABRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060491.1 | XP_017915980.1 | 3 - 4 | 379 | 59 | |
| GenBank mRNA | JO630542.2 | - | 6 - 7 | 1515 | 59 | |
More Information
Set Membership: |
|

GABRD
Genomic Map