Assay Details
| Gene Symbol: GLIS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044355.1 | XP_017899844.1 | 7 - 8 | 1817 | 68 | |
| GenBank mRNA | JO454619.2 | - | 3 - 4 | 337 | 68 | |
More Information
Set Membership: |
|

GLIS1
Genomic Map