Assay Details
Gene Symbol: MTHFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055019.1 | XP_017910508.1 | 7 - 8 | 959 | 79 | |
XM_018055020.1 | XP_017910509.1 | 6 - 7 | 836 | 79 | ||
GenBank mRNA | JO586833.2 | - | 1 - 2 | 1236 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
