Assay Details
| Gene Symbol: CNEP1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061953.1 | XP_017917442.1 | 5 - 6 | 465 | 83 | |
| GenBank mRNA | JO363181.2 | - | 5 - 6 | 414 | 83 | |
| KA333745.1 | - | 1 - 2 | 580 | 83 | ||
More Information
Set Membership: |
|

CNEP1R1
Genomic Map