Assay Details
Gene Symbol: RBAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697901.3 | XP_005697958.1 | 1 - 2 | 1107 | 105 | |
XM_005697903.3 | XP_005697960.1 | 3 - 4 | 693 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
