Assay Details
Gene Symbol: GPBAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059576.1 | XP_017915065.1 | 2 | 1320 | 85 | |
XM_018059581.1 | XP_017915070.1 | 3 | 1413 | 85 | ||
GenBank mRNA | JO416129.2 | - | 2 | 1087 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
