Assay Details
| Gene Symbol: DNAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754093.4 | XP_002754139.1 | 5 - 6 | 341 | 119 | |
| XM_009006314.2 | XP_009004562.1 | 5 - 6 | 371 | 119 | ||
More Information
Set Membership: |
|

DNAL1
Genomic Map