Assay Details
| Gene Symbol: ZPLD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965278.1 | XP_017820767.1 | 4 - 5 | 977 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZPLD1| Gene Symbol: ZPLD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965278.1 | XP_017820767.1 | 4 - 5 | 977 | 118 | |
Set Membership: |
|