Assay Details
| Gene Symbol: GLDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753484.3 | XP_002753530.1 | 4 - 5 | 703 | 86 | |
| XM_009005450.2 | XP_009003698.1 | 4 - 5 | 589 | 86 | ||
More Information
Set Membership: |
|

GLDN
Genomic Map