Assay Details
| Gene Symbol: FAM222A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752983.3 | XP_002753029.1 | 2 - 3 | 791 | 73 | |
| XM_017976366.1 | XP_017831855.1 | 2 - 3 | 212 | 73 | ||
More Information
Set Membership: |
|

FAM222A
Genomic Map