Assay Details
| Gene Symbol: HSDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761227.3 | XP_002761273.1 | 4 - 5 | 847 | 63 | |
| XM_017966839.1 | XP_017822328.1 | 4 - 5 | 1471 | 63 | ||
More Information
Set Membership: |
|

HSDL1
Genomic Map