Assay Details
| Gene Symbol: CELF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753307.3 | XP_002753353.1 | 3 - 4 | 836 | 60 | |
| XM_003733912.3 | XP_003733960.1 | 3 - 4 | 836 | 60 | ||
| XM_009005192.2 | XP_009003440.1 | 3 - 4 | 836 | 60 | ||
| XM_009005193.2 | XP_009003441.1 | 3 - 4 | 836 | 60 | ||
| XM_009005194.2 | XP_009003442.1 | 3 - 4 | 836 | 60 | ||
| XM_009005195.2 | XP_009003443.1 | 3 - 4 | 836 | 60 | ||
| XM_009005196.2 | XP_009003444.1 | 3 - 4 | 836 | 60 | ||
| XM_009005198.2 | XP_009003446.1 | 2 - 3 | 241 | 60 | ||
| XM_017976742.1 | XP_017832231.1 | 3 - 4 | 836 | 60 | ||
More Information
Set Membership: |
|

CELF6
Genomic Map