Assay Details
| Gene Symbol: GLRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745204.3 | XP_002745250.1 | 6 - 7 | 1151 | 103 | |
| XM_002745205.3 | XP_002745251.1 | 6 - 7 | 1151 | 103 | ||
More Information
Set Membership: |
|

GLRA3
Genomic Map