Assay Details
| Gene Symbol: ETV5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981351.2 | XP_008979599.1 | 8 - 9 | 1159 | 89 | |
| XM_017964741.1 | XP_017820230.1 | 8 - 9 | 1047 | 89 | ||
More Information
Set Membership: |
|

ETV5
Genomic Map