Assay Details
| Gene Symbol: CHMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757526.4 | XP_002757572.1 | 5 - 6 | 698 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHMP3| Gene Symbol: CHMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757526.4 | XP_002757572.1 | 5 - 6 | 698 | 86 | |
Set Membership: |
|