Assay Details
| Gene Symbol: ARHGEF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989390.2 | XP_008987638.2 | 3 - 4 | 956 | 60 | |
| XM_008989394.2 | XP_008987642.2 | 2 - 3 | 1216 | 60 | ||
| XM_017968513.1 | XP_017824002.1 | 2 - 3 | 1212 | 60 | ||
More Information
Set Membership: |
|

ARHGEF9
Genomic Map