Assay Details
| Gene Symbol: PATZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009484.2 | XP_009007732.2 | 6 - 7 | 2118 | 63 | |
| XM_017979258.1 | XP_017834747.1 | 6 - 7 | 2183 | 63 | ||
More Information
Set Membership: |
|

PATZ1
Genomic Map