Assay Details
| Gene Symbol: TMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997839.2 | XP_008996087.1 | 9 - 10 | 2260 | 65 | |
| XM_017972885.1 | XP_017828374.1 | 10 - 11 | 2482 | 65 | ||
| XM_017972886.1 | XP_017828375.1 | 8 - 9 | 2366 | 65 | ||
| XM_017972887.1 | XP_017828376.1 | 8 - 9 | 2366 | 65 | ||
More Information
Set Membership: |
|

TMC8
Genomic Map