Assay Details
| Gene Symbol: PCGF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756569.3 | XP_002756615.1 | 4 - 5 | 689 | 119 | |
| XM_009010217.2 | XP_009008465.1 | 4 - 5 | 689 | 119 | ||
| XM_017979279.1 | XP_017834768.1 | 4 - 5 | 689 | 119 | ||
| XM_017979280.1 | XP_017834769.1 | 4 - 5 | 689 | 119 | ||
More Information
Set Membership: |
|

PCGF6
Genomic Map