Assay Details
| Gene Symbol: CSRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752804.4 | XP_002752850.1 | 2 - 3 | 237 | 82 | |
| XM_009004268.1 | XP_009002516.1 | 2 - 3 | 158 | 82 | ||
| XM_009004269.2 | XP_009002517.1 | 2 - 3 | 230 | 82 | ||
| XM_009004270.2 | XP_009002518.1 | 1 - 2 | 127 | 82 | ||
More Information
Set Membership: |
|

CSRP2
Genomic Map