Assay Details
| Gene Symbol: BCL9L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977595.1 | XP_017833084.1 | 5 - 6 | 778 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BCL9L| Gene Symbol: BCL9L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977595.1 | XP_017833084.1 | 5 - 6 | 778 | 58 | |
Set Membership: |
|