Assay Details
| Gene Symbol: MAT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756181.3 | XP_002756227.1 | 9 - 10 | 1651 | 82 | |
| XM_009009649.2 | XP_009007897.1 | 9 - 10 | 1673 | 82 | ||
More Information
Set Membership: |
|

MAT1A
Genomic Map