Assay Details
| Gene Symbol: TMEM248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743903.3 | XP_002743949.1 | 5 - 6 | 1112 | 105 | |
| XM_017965902.1 | XP_017821391.1 | 5 - 6 | 1107 | 105 | ||
| XM_017965908.1 | XP_017821397.1 | 5 - 6 | 1081 | 105 | ||
| XM_017965913.1 | XP_017821402.1 | 5 - 6 | 902 | 105 | ||
More Information
Set Membership: |
|

TMEM248
Genomic Map