Assay Details
| Gene Symbol: NEGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750998.4 | XP_002751044.1 | 1 - 2 | 370 | 77 | |
| XM_017974842.1 | XP_017830331.1 | 1 - 2 | 370 | 77 | ||
More Information
Set Membership: |
|

NEGR1
Genomic Map