Assay Details
| Gene Symbol: DCLRE1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756605.4 | XP_002756651.1 | 5 - 6 | 3148 | 101 | |
| XM_009010308.2 | XP_009008556.1 | 5 - 6 | 3148 | 101 | ||
| XM_009010309.2 | XP_009008557.1 | 5 - 6 | 3148 | 101 | ||
More Information
Set Membership: |
|

DCLRE1A
Genomic Map