Assay Details
| Gene Symbol: EMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757630.4 | XP_002757676.1 | 3 - 4 | 1064 | 82 | |
| XM_008980516.2 | XP_008978764.1 | 2 - 3 | 430 | 82 | ||
More Information
Set Membership: |
|

EMX1
Genomic Map