Assay Details
| Gene Symbol: ABHD17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742853.3 | XP_002742899.1 | 2 - 3 | 1076 | 101 | |
| XM_008995644.2 | XP_008993892.1 | 2 - 3 | 596 | 101 | ||
More Information
Set Membership: |
|

ABHD17B
Genomic Map