Assay Details
| Gene Symbol: LRRC8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001654.2 | XP_008999902.1 | 5 - 6 | 2545 | 89 | |
| XM_017974909.1 | XP_017830398.1 | 4 - 5 | 4603 | 89 | ||
| XM_017974910.1 | XP_017830399.1 | 5 - 6 | 3326 | 89 | ||
| XM_017974911.1 | XP_017830400.1 | 6 - 7 | 2646 | 89 | ||
More Information
Set Membership: |
|

LRRC8B
Genomic Map