Assay Details
| Gene Symbol: UHRF1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746456.3 | XP_002746502.1 | 17 - 18 | 3952 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UHRF1BP1| Gene Symbol: UHRF1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746456.3 | XP_002746502.1 | 17 - 18 | 3952 | 75 | |
Set Membership: |
|