Assay Details
| Gene Symbol: SPRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757686.4 | XP_002757732.1 | 6 - 7 | 1018 | 73 | |
| XM_002757687.4 | XP_002757733.1 | 5 - 6 | 1471 | 73 | ||
More Information
Set Membership: |
|

SPRED2
Genomic Map