Assay Details
| Gene Symbol: UBTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756492.3 | XP_002756538.1 | 2 - 3 | 841 | 73 | |
| XM_009010095.2 | XP_009008343.2 | 2 - 3 | 413 | 73 | ||
| XM_017979208.1 | XP_017834697.1 | 2 - 3 | 956 | 73 | ||
More Information
Set Membership: |
|

UBTD1
Genomic Map