Assay Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762068.3 | XP_002762114.1 | 1 - 2 | 174 | 105 | |
| XM_003735561.3 | XP_003735609.1 | 1 - 2 | 159 | 105 | ||
| XM_008987899.2 | XP_008986147.1 | 1 - 2 | 107 | 105 | ||
| XM_008987900.2 | XP_008986148.1 | 1 - 2 | 107 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002762068.3, XM_003735561.3, XM_008987899.2, XM_008987900.2 transcripts.
Set Membership: |
|

ZNF527
Genomic Map