Assay Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762068.3 | XP_002762114.1 | 4 - 5 | 469 | 74 | |
| XM_008987899.2 | XP_008986147.1 | 4 - 5 | 392 | 74 | ||
| XM_017968136.1 | XP_017823625.1 | 3 - 4 | 263 | 74 | ||
More Information
Set Membership: |
|

ZNF527
Genomic Map