Assay Details
| Gene Symbol: SGCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988730.2 | XP_008986978.1 | 4 - 5 | 816 | 67 | |
| XM_008988734.2 | XP_008986982.1 | 4 - 5 | 397 | 67 | ||
| XM_008988740.2 | XP_008986988.1 | 3 - 4 | 949 | 67 | ||
| XM_008988745.2 | XP_008986993.1 | 4 - 5 | 799 | 67 | ||
| XM_008988748.2 | XP_008986996.1 | 3 - 4 | 380 | 67 | ||
| XM_017969165.1 | XP_017824654.1 | 4 - 5 | 581 | 67 | ||
| XM_017969169.1 | XP_017824658.1 | 4 - 5 | 816 | 67 | ||
More Information
Set Membership: |
|

SGCD
Genomic Map