Assay Details
| Gene Symbol: CCDC59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752795.4 | XP_002752841.1 | 3 - 4 | 695 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC59| Gene Symbol: CCDC59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752795.4 | XP_002752841.1 | 3 - 4 | 695 | 65 | |
Set Membership: |
|