Assay Details
| Gene Symbol: AKAP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763215.3 | XP_002763261.1 | 4 - 5 | 547 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AKAP14| Gene Symbol: AKAP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763215.3 | XP_002763261.1 | 4 - 5 | 547 | 75 | |
Set Membership: |
|