Assay Details
| Gene Symbol: FCGR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759850.4 | XP_002759896.1 | 5 - 6 | 929 | 67 | |
| XM_008984246.2 | XP_008982494.1 | 6 - 7 | 896 | 67 | ||
| XM_017966223.1 | XP_017821712.1 | 4 - 5 | 904 | 67 | ||
More Information
Set Membership: |
|

FCGR1A
Genomic Map