Assay Details
| Gene Symbol: SEC61G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009002189.2 | XP_009000437.1 | 2 - 3 | 264 | 95 | |
| XM_009002190.2 | XP_009000438.1 | 2 - 3 | 187 | 95 | ||
| XM_009002191.2 | XP_009000439.1 | 2 - 3 | 188 | 95 | ||
| XM_009002192.1 | XP_009000440.1 | 2 - 3 | 180 | 95 | ||
More Information
Set Membership: |
|

SEC61G
Genomic Map