Assay Details
| Gene Symbol: GABRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009000274.1 | XP_008998522.1 | 8 | 1388 | 93 | |
| GenBank mRNA | EF214950.1 | - | 1 | 217 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GABRD| Gene Symbol: GABRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009000274.1 | XP_008998522.1 | 8 | 1388 | 93 | |
| GenBank mRNA | EF214950.1 | - | 1 | 217 | 93 | |
Set Membership: |
|